Powder for dish washers, neodisher® MA
Catalog # 148-3132
Supplier: Dr. Weigert
Specifications
- Description:neodisher® MA*
- Pk:10 kg
Specifications
About this item
Mildly alkaline detergent for washing machines with applications including cleaning of surgical instruments, anaesthesia materials, operating theatre clogs and glassware used in medical, biological and chemical laboratories.
- Free from oxidising materials
- Very good cleaning action and very gentle on material
- Mildly alkaline formulation with defoaming properties
- Phosphates>30%
- Non-ionic surfactants<5%
- Dosage: Typically 1 to 4 g/litre