WHEATON® Bottle, Wide Neck, Round, AC Type
Supplier: DWK Life Sciences
Certificates
About this item
Bottles manufactured from clear soda lime glass.
- Packaged in convenience packs with caps attached to maintain cleanliness
- Hazardous communication label included with each case of bottles which allows for content ID and safety notification
- Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Wide mouth bottles are ideal for dry and viscous samples.