Order Entry
Finland
ContactUsLinkComponent
Weighing dishes, aluminium
Weighing dishes, aluminium
Catalog # HEATHS14522
Supplier:  HEATHROW
undefined
Weighing dishes, aluminium
Catalog # HEATHS14522
Supplier:  HEATHROW
Specifications
  • Ext.Ø:
    101 mm
  • Height:
    8 mm
  • Pk:
    50
Specifications
About this item
Weighing dishes, made from light gauge aluminium are designed for moisture determination balances. Ideal for general-purpose weighing and dispensing.
  • Used as evaporating dish or dust cover
  • Lightweight, sturdy dish has no coating and is oil free
  • Slightly tapered for stacking