3-Methoxycyclohex-2-en-1-one for synthesis
Catalog # 8.14561.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:32 - 35 °C
- Molar Mass:126.16 g/mol
- Boiling Point:114 °C (19 hPa)
Specification Test Results
About this item
:
:126.16 g/mol
:M-Clarity™ quality level = MQ200
EINECS:240-837-0
CAS Number:16807-60-6
Formula:C₇H₁₀O₂
MDL Number:MFCD00236366