Order Entry
Finland
ContactUsLinkComponent
Pyromellitic dianhydride for synthesis, Sigma-Aldrich®
Health
Corrosive
Pyromellitic dianhydride for synthesis, Sigma-Aldrich®
Catalog # 8.07516.0100
Supplier:  Merck
CAS Number:  
Health
Corrosive
Pyromellitic dianhydride for synthesis, Sigma-Aldrich®
Catalog # 8.07516.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :218.12 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:201-898-9
 :GHS08
 :GHS05
Storage Temperature:Ambient
CAS Number:89-32-7
Formula:C₁₀H₂O₆
MDL Number:MFCD00005005