Order Entry
Finland
ContactUsLinkComponent
Aurintricarboxylic acid, Millipore®
Aurintricarboxylic acid, Millipore®
Catalog # 189400-100
CAS Number:  
Aurintricarboxylic acid, Millipore®
Catalog # 189400-100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :422.35 g/mol
EINECS:224-628-1
CAS Number:4431-00-9
Formula:C₂₂H₁₄O₉
MDL Number:MFCD00011663