Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aurintricarboxylic acid
Aurintricarboxylic acid
Aurintricarboxylic acid
:
Thermo Fisher Scientific
Aurintricarboxylic acid
:
Thermo Fisher Scientific
:
4431-00-9
:
:
C₂₂H₁₄O₉
:
MFCD00011663
:
422.35 g/mol
:
>300 °C
:
224-628-1
:
GHS07
:
Ambient
msds_ALFAA15905
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years