Funnels with long stem, BOLA
Supplier: Bohlender
Certificates
About this item
Funnels, made of PTFE, white. Conical opening with long outlet.
- Temperature resistance from −200 to +250 °C
- Universal chemical resistance
Funnels, made of PTFE, white. Conical opening with long outlet.