Distillation apparatus, flat flange, BOLA
Supplier: Bohlender
Certificates
About this item
PTFE/PFA.
- Temperature resistance: from −200 to +200 °C
- Universal chemical resistance
- Vacuum: suitable
Suitable for the distillation of strong alkaline or acid products as well as very aggressive solvents.