Test strips, blank, Reflectoquant®, Supelco®
Catalog # 1.16730.0001
Supplier: Merck
CAS Number:
Specifications
- Pk:50 Tests
- Description:Blank strip (colour correction)
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly