VWR®, Filter Tips
Transparent PP, sterile.
- E-beam sterilised tips are free from DNA, DNase and RNase, pyrogens, PCR inhibitors and endotoxins
- Robust, traceable racks are easy to grab with their ergonomic clips
- Reload boxes (including racks) feature a flip-lock latch for one handed opening and closing, reversible lid with lip, flush fitting and click-clip to stabilise the racks
: SC: Single-channel pipette / MC: Multi-channel pipette
For more compatibility information, please contact Avantor Customer Services.
For more compatibility information, please contact Avantor Customer Services.