Acridine yellow G ≥90% (dye basis), Sigma-Aldrich®
Catalog # 199508-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:273.76 g/mol
:M-Clarity™ quality level = MQ100
EINECS:205-194-2
:GHS07
:GHS08
CAS Number:135-49-9
Formula:C₁₅H₁₅N₃·HCl
MDL Number:MFCD00012661