Order Entry
Finland
ContactUsLinkComponent
2-Bromo-5-methylpyridine 98%
Irritant
2-Bromo-5-methylpyridine 98%
Certificates
undefined
Irritant
2-Bromo-5-methylpyridine 98%

Specification Test Results

  • Appearance:
    White to light yellow to light brown Low melting crystalline solid or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    40 to 45 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information