Order Entry
Finland
ContactUsLinkComponent
 
 
2-Chloro-6-methoxy-3-nitropyridine ≥98.0% (by GC)
 :  TCI
 
undefined
 
2-Chloro-6-methoxy-3-nitropyridine ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    76.0 to 80.0 deg-C