Industrial gloves, Polytril™
Supplier: HONEYWELL SAFETY
Certificates
About this item
White polyamide knitted glove with grey oilproof nitrile coating on palm and fingertips and elastic knitted wrist.
- Tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The ventilated back improves the glove's breathability
- Knitted and elasticated wrist gives the glove good support
- Nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
Suitable for handling of fine objects in a greasy, damp or soiled atmosphere and mechanical handling in the automotive industry.
Certifications: EN 388:2016 (4121X); Cat. II.
Specifications
- Catalog No:
- 112-1518
- 112-1517
- 112-1523
- 112-1519
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Finish:
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)