2-Naphthol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:135-19-3
:
Formula:C₁₀H₈O
MDL Number:MFCD00004067
Merck Index:12,06471
Boiling Pt:285…286 °C (760 mmHg)
Melting Pt:120…124 °C
Density:1,217 g/cm³ (20 °C)
Flash Pt:153 °C
UN:3077
ADR:9,III
:144.17 g/mol
EINECS:205-182-7
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige to pink to light brown Powder or flakes
- Infrared spectrum:Conforms
- Melting point:119 to 125 °C
- GC:≥97.5 %