Silica gel, Pore size: 60 Å
Catalog # MOLE22768261-250G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:60.08 g/mol
Storage Temperature:Ambient
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232