Silica gel, Pore size: 40 Å 0.200-0.500 mm for chromatography
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7631-86-9
:112926-00-8
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1710 °C
Storage Temperature:Ambient
REACH:01-2119379499-16
Specification Test Results
- Appearance:White Crystals
- Particle size:0.2 to 0.5 mm
- Specific gravity:0.54 to 0.6 kg/l
- pH:5.5 to 7 (10 % suspension in water)
- Residual water:2 to 4 % (160 °C)