Order Entry
Finland
ContactUsLinkComponent
Ethylenediammonium sulphate ≥97% for synthesis, Sigma-Aldrich®
Ethylenediammonium sulphate ≥97% for synthesis, Sigma-Aldrich®
Catalog # 8.20031.0250
Supplier:  Merck
CAS Number:  
Ethylenediammonium sulphate ≥97% for synthesis, Sigma-Aldrich®
Catalog # 8.20031.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    225 - 228 °C
  • Molar Mass:
    158.17 g/mol

Specification Test Results

About this item

 :
 :158.17 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:206-369-6
CAS Number:22029-36-3
Formula:C₂H₈N₂
MDL Number:MFCD00036417