Glyoxylic acid monohydrate, Sigma-Aldrich®
: Merck
:563-96-2
:MFCD00127974
:C₂H₂O₃·1H₂O
:MFCD00149497
:92.05 g/mol
:8,III
:M-Clarity™ quality level = MQ200
:206-058-5
:3261
:GHS05
:GHS07
:Ambient
- Solubility:(20 °C) soluble
- Melting Point:46.7 °C
- Molar Mass:92.05 g/mol
- Hazard Class:8
- Hazard Class Description:Corrosive Material