Methacrylonitrile
Glass bottle, 250 mg
Catalog # EHERCA14972000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- CAS Number:126-98-7
- Formula:C₄H₅N
- Molecular Weight:67.09 g/mol
- EINECS:204-817-5
- UN:3079
- ADR:3,I
Specifications
Specifications
- Pack type:Glass bottle
- Pk:250 mg



