Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Strontium nitrate
Strontium nitrate ≥99.999% (metals basis), Puratronic®
Strontium nitrate ≥99.999% (metals basis), Puratronic®
:
Thermo Fisher Scientific
Strontium nitrate ≥99.999% (metals basis), Puratronic®
:
Thermo Fisher Scientific
:
10042-76-9
:
:
Sr(NO₃)₂
:
MFCD00011248
:
211.63 g/mol
:
14,8845
:
645 °C (1013 hPa)
:
570 °C
:
2.990
:
1507
:
5.1,III
:
Packaged under argon.
:
233-131-9
:
GHS03
:
GHS07
:
Ambient
msds_ALFA043193