
Strontium chloride, anhydrous 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10476-85-4
:
Formula:Cl₂Sr
MDL Number:MFCD00011249
:158.53 g/mol
Merck Index:12,09000
Boiling Pt:1250 °C (1013 hPa)
Melting Pt:873 °C
Density:1,96 g/cm³ (20 °C)
EINECS:233-971-6
:GHS05
Specification Test Results
- Appearance:White Crystalline powder
- Assay:54.4 to 55.9 % (Sr)
- Total trace metal impurities:≤0.01 %