Sodium metabisulphite
Catalog # MOLE33231649-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:190.11 g/mol
Merck Index:12,08784
Melting Pt:150 °C
Density:1.48 g/cm³ (20 °C)
EINECS:231-673-0
REACH:01-2119531326-45
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7681-57-4
Formula:Na₂S₂O₅
MDL Number:MFCD00167602