Sodium metabisulphite ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7681-57-4
:
Formula:Na₂S₂O₅
MDL Number:MFCD00167602
:190.11 g/mol
Merck Index:14,8638
Melting Pt:>300 °C
Density:1.480
EINECS:231-673-0
REACH:01-2119531326-45
:GHS05
:GHS07
Storage Temperature:Ambient