Order Entry
Finland
Orders LinkContactUsLinkComponent
Sodium thiosulphate

Sodium thiosulphate

Glass bottle for solids, 500 g
Catalog # MOLE43109159-500G
Marketsource
Supplier:   Molekula
Sodium thiosulphate
Catalog # MOLE43109159-500G
Supplier:  Molekula
Marketsource
CAS Number:  7772-98-7
Sodium thiosulphate

Glass bottle for solids, 500 g

CAS Number:  7772-98-7
Price
61,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  • CAS Number:7772-98-7
  • Formula:Na₂S₂O₃
  • MDL Number:MFCD00003499
  • Molecular Weight:158.11 g/mol
  • Merck Index:12,08844
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:48 °C
  • Density:1.66 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-867-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.