Sodium thiosulphate, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7772-98-7
:
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
:158.11 g/mol
Merck Index:12,08844
Boiling Pt:100 °C (1013 hPa)
Melting Pt:48 °C
Density:1.66 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-867-5
Related Information
Certificates
Search by Lot NumberMERC1.06512.2500_TDS_EN ProductPage_106512
Product specification
msds_106512