Order Entry
Finland
ContactUsLinkComponent
Sodium hydroxide ≥98.0% (by titrimetric analysis), pellets, Puriss. p.a. ACS (max. 0.02% K), Fluka™
Sodium hydroxide ≥98.0% (by titrimetric analysis), pellets, Puriss. p.a. ACS (max. 0.02% K), Fluka™
Certificates
Sodium hydroxide ≥98.0% (by titrimetric analysis), pellets, Puriss. p.a. ACS (max. 0.02% K), Fluka™
Certificates

Specification Test Results

  • Assay:
    Min. 98.0 %
  • Assay of Na₂CO₃:
    Max. 1.0 %
  • Silver (Ag):
    Max. 0.0005 %
  • Aluminium (Al):
    Max. 0.0005 %
  • Barium (Ba):
    Max. 0.0005 %
  • Bismuth (Bi):
    Max. 0.0005 %
  • Calcium (Ca):
    Max. 0.0005 %
  • Cadmium (Cd):
    Max. 0.0005 %
  • Cobalt (Co):
    Max. 0.0005 %
  • Chromium (Cr):
    Max. 0.0005 %
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 0.0005 %
  • Mercury (Hg):
    Max. 0.00001 %
  • Potassium (K):
    Max. 0.02 %
  • Lithium (Li):
    Max. 0.0005 %
  • Magnesium (Mg):
    Max. 0.0005 %
  • Manganese (Mn):
    Max. 0.0005 %
  • Molybdenum (Mo):
    Max. 0.0005 %
  • Nickel (Ni):
    Max. 0.0005 %
  • Lead (Pb):
    Max. 0.0005 %
  • Strontium (Sr):
    Max. 0.0005 %
  • Thallium (Tl):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.0005 %
  • Heavy metals (as Ag):
    Max. 0.002 %
  • Chloride (Cl):
    Max. 0.005 %
  • Total N:
    Max. 0.0003 %
  • Phosphate (PO₄):
    Max. 0.0005 %
  • Silicate (as SiO₂):
    Max. 0.0005 %
  • Sulfate (SO₄):
    Max. 0.003 %
  • Vapor Pressure:
    3 mmHg ( 37 °C)

Specification Test Results

Related Information