Sodium hydroxide ≥98%, pellets, EMPROVE® ESSENTIAL Ph. Eur., BP, FCC, JP, NF, E524, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
:GHS05
Specification Test Results
- Assay (acidimetric,NaOH):≥ 98.0 %
- Assay (total alkalinity calc. as NaOH):98.0 - 100.5 %
- Assay (Na):54.0 - 59.8 %
- Identity:passes test
- Appearance of solution:passes test
- Insoluble substances and organic matter:passes test
- Carbonate (as Na₂CO₃):≤ 0.5 %
- Chloride (Cl):≤ 0.015 %
- Phosphate (PO₄):≤ 0.002 %
- Silicate (SiO₂):≤ 0.01 %
- Sulfate (SO₄):≤ 0.010 %
- Total nitrogen (N):≤ 0.0005 %
- Heavy metals (as Pb):≤ 0.0005 %
- Al (Aluminium):≤ 0.001 %
- As (Arsenic):≤ 0.0003 %
- Cu (Copper):≤ 0.0005 %
- Fe (Iron):≤ 0.0010 %
- Hg (Mercury):≤ 0.00001 %
- K (Potassium):≤ 0.1
- Pb (Lead):≤ 0.00005 %
- Zn (Zinc):≤ 0.0025 %
- Residual solvents (Ph Eur/USP/ICH):excluded by manufacturing process
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERP1.06482.1000_TDS_EN MERP1.06482.5000_TDS_EN ProductPage_106482
Product specification
msds_106482