Sodium hydroxide, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
UN:1823
ADR:8,II
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:215-185-5
REACH:01-2119457892-27
:GHS05
Specification Test Results
- Solubility:1090 g/l (20 °C)
- Melting Point:323 °C
- Molar Mass:40.00 g/mol
- Boiling Point:1390 °C (1013 hPa)
- Vapor Pressure:(20 °C)
- Density:2.13 g/cm3 (20 °C)
- pH:14 (50 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_106467
SDS
ProductPage_106467 ProductPage_106467