Sodium hydroxide, beads, Reagent Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
:GHS05
Specification Test Results
- Chloride:≤0.05%
- Heavy Metals:≤0.006%
- Potassium:≤0.1%
- Purity:>98.5%
- Sodium Carbonate:≤0.90%
- Sulphate:≤0.005%