Potassium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7757-79-1
:
Formula:KNO₃
MDL Number:MFCD00011409
:101.1 g/mol
Merck Index:13,07733
Melting Pt:334 °C
Density:2.109 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-818-8
UN:1486
ADR:5.1,III
REACH:01-2119488224-35
:GHS03
Specification Test Results
- Assay:Min. 99.0 %
- pH (20°C; 5 %):5.0 - 7.5
- Heavy metals (as Pb):Max. 5 ppm
- Cl (Chloride):Max. 10 ppm
- IO3 (Iodate):Max. 5 ppm
- NH4 (Ammonium):Max. 10 ppm
- NO2 (Nitrite):Max. 10 ppm
- PO4 (Phosphate):Max. 5 ppm
- SO4 (Sulphate):Max. 30 ppm
- Ca (Calcium):Max. 10 ppm
- Cu (Copper):Max. 1 ppm
- Fe (Iron):Max. 3 ppm
- Mg (Magnesium):Max. 15 ppm
- Na (Sodium):Max. 0.02 %
- Pb (Lead):Max. 1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
26869_POTASSIUM NITRATE ANALAR NORMAPUR_BSE_REV0.pdf
BSE/TSE Certificates
spec_VWRC26869.234 spec_VWRC26869.234 spec_VWRC26869.260 spec_VWRC26869.260 spec_VWRC26869.291
Product specification
More