Order Entry
Finland
ContactUsLinkComponent
Potassium nitrate ≥99.0%, crystalline powder ACS
Potassium nitrate ≥99.0%, crystalline powder ACS
Supplier:  MP Biomedicals
Certificates
Potassium nitrate ≥99.0%, crystalline powder ACS
Supplier:  MP Biomedicals

Specification Test Results

  • Synonyms:
    Niter; Nitre; Nitric Acid Potassium Salt; Saltpeter; Vicknite
  • Form:
    White Crystalline Powder
  • Storage:
    Store at room temperature (15 to 30 °C)
  • Assay:
    ≥99,0%
  • Heavy Metals:
    ≤5 ppm (as Pb); Iron (Fe): ≤3 ppm
  • Insoluble Matter:
    ≤0,005%
  • pH:
    4,5 - 8,5 (5% aq soln)

Specification Test Results