Potassium fluoride
Glass bottle for solids, 125 g
Catalog # PROONIST-2203
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:7789-23-3
- Formula:KF
- MDL Number:MFCD00011398
- Molecular Weight:58.1 g/mol
- Merck Index:13,07714
- Boiling Pt:1505 °C (1013 hPa)
- Melting Pt:860 °C
- Density:2.48 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-151-5
- UN:1812
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:125 g



