Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Potassium dichromate

Oxidizing
Toxic
Corrosive
Health
Environment
Marketsource
Supplier:   Roth Carl
Potassium dichromate
Supplier:  Roth Carl
Marketsource
Certificates
Potassium dichromate

2 Options Available Below

Product Details & Documents
  • CAS Number:7778-50-9
  • Formula:K₂Cr₂O₇
  • MDL Number:MFCD00011367
  • Molecular Weight:294.18 g/mol
  • Merck Index:12,07790
  • Boiling Pt:500 °C (1013 hPa)
  • Melting Pt:398 °C
  • Density:2.67 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-906-6
  • UN:3087
  • ADR:5.1,II
  • REACH:01-2119454792-32
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 260,59 €
    260,59 €
    1 * 1 kg
       260,59 €

    Specifications:
    Marketsource
    Marketsource
  • Plastic bottle for solids
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 1 028,97 €
    1 028,97 €
    1 * 5 kg
       1 028,97 €

    Specifications:
    Marketsource
    Marketsource