Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7447-40-7
:
Formula:KCl
MDL Number:MFCD00011360
:74.55 g/mol
Merck Index:14,07621
Boiling Pt:1420 °C (1013 hPa)
Melting Pt:773 °C
Density:1.98 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:231-211-8
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:347 g/l (20 °C)
- Melting Point:773 °C
- Molar Mass:74.55 g/mol
- Bulk Density:1000 kg/m3
- Boiling Point:1413 °C (1013 hPa)
- Density:1.98 g/cm3 (20 °C)
- pH:5.5 - 8.0 (50 g/l, H2O, 25 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Individual products and tailor-made chemicals for your technical applications
Product Literature
msds_104936
SDS
ProductPage_104936