Cesium chloride
Catalog # MOLEM82986284
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:168.36 g/mol
Merck Index:13,02021
Boiling Pt:1303 °C (1013 hPa)
Melting Pt:646 °C
Density:3.7808 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-600-2
CAS Number:7647-17-8
Formula:CsCl
MDL Number:MFCD00010955