Cesium chloride, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-17-8
:
Formula:CsCl
MDL Number:MFCD00010955
:168.36 g/mol
Merck Index:14,2011
Boiling Pt:1303 °C
Melting Pt:646 °C
Density:3.99
:ampouled under argon
Storage Temperature:Ambient
EINECS:231-600-2