Order Entry
Finland
ContactUsLinkComponent
Sodium hydrogen carbonate ≥99.7%, powder, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Sodium hydrogen carbonate ≥99.7%, powder, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Certificates
undefined
Sodium hydrogen carbonate ≥99.7%, powder, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Certificates

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Assay (excluding dried substance):
    99.7 - 100.3 %
  • Water insoluble matter:
    Max. 0.015 %
  • pH (5 %, 25 °C):
    Max. 8.6
  • Ammonium (NH₄):
    Max. 5 ppm
  • Arsenic (As):
    Max. 2 ppm
  • Calcium (Ca):
    Max. 100 ppm
  • Cadmium (Cd):
    Max. 0.0005 %
  • Cobalt (Co):
    Max. 50 ppm
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Nickel (Ni):
    Max. 50 ppm
  • Lead (Pb):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 30 ppm
  • Phosphate (PO₄):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 150 ppm
  • I reducing matters (as SO₂):
    Max. 0.005 %
  • S-compounds (as SO₄):
    Max. 0.003 %
  • Colour of the solution:
    complying
  • Turbidity of solution:
    complying
  • Solution 0.5 g at 10 mL (H₂O):
    complying
  • Residual solvents:
    complying

Specification Test Results

Related Information