Tubing connectors, straight
Supplier: KARTELL
Certificates
About this item
PP, white.
- Good mechanical strength and chemical resistance
- Excellent thermal resistance, autoclavable
- Conform to foodstuff regulations 90/128/CEE, 92/39/CEE and FDA 21 CFR
With tapered ends, for connecting tubing with different inner diameters.