Order Entry
Finland
Orders LinkContactUsLinkComponent
Potassium bromate ≥99%

Potassium bromate ≥99%

Oxidizing
Toxic
Health
Potassium bromate ≥99%
Certificates
Potassium bromate ≥99%
Potassium bromate ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 7758-01-2 Molecular Formula: KBrO3 Formula Weight: 167.00 Odour: Odourless Melting Point: ca 370° dec. UN No.: 1484 MDL No.: MFCD00011359

  • CAS Number:7758-01-2
  • Formula:KBrO₃
  • MDL Number:MFCD00011359
  • Molecular Weight:167 g/mol
  • Merck Index:14,7617
  • Melting Pt:∼ 370 °C (dec.)
  • Density:3.27
  • Storage Temperature:Ambient
  • EINECS:231-829-8
  • UN:1484
  • ADR:5.1,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 49,20 €
    49,20 €
    1 * 100 g
       49,20 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 126,00 €
    126,00 €
    1 * 500 g
       126,00 €

  • Glass bottle for solids
    2.500 g
    Log in for availability
    Log in for availability
    1 * 2.500 g
    1 * 2.500 g - 476,00 €
    476,00 €
    1 * 2.500 g
       476,00 €