Lithium bromide, anhydrous 99+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7550-35-8
:23303-71-1
Formula:LiBr
MDL Number:MFCD00011077
:86.84 g/mol
Merck Index:13,05548
Boiling Pt:1265 °C (760 mmHg)
Melting Pt:550 °C
Density:3.46 g/cm³
EINECS:231-439-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration Argentometric:≥99.0 %
- Loss on drying:≤0.5 % (400°C)
- pH:6 to 12 (4 % at 20°C)
- Chloride (Cl):≤1500 ppm
- Carbonate (CO3):≤300 ppm
- Sulfate (SO4):≤200 ppm
- Calcium (Ca):≤50 ppm
- Copper (Cu):≤3 ppm
- Iron (Fe):≤10 ppm
- Magnesium (Mg):≤20 ppm
- Potassium (K):≤300 ppm
- Sodium (Na):≤400 ppm
- Insoluble matter:≤0.005 % (in water)