Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standard, 6-Nitrochrysene 50 µg/ml in toluene, CIL
Organic reference standard, 6-Nitrochrysene 50 µg/ml in toluene, CIL
Organic reference standard, 6-Nitrochrysene 50 µg/ml in toluene, CIL
PROOCILULM38811.2
:
LGC Standards PROMOCHEM
:
Organic reference standard, 6-Nitrochrysene 50 µg/ml in toluene, CIL
PROOCILULM38811.2
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,2 mL
CAS No.:
7496-02-8
Description:
6-Nitrochrysene 50 µg/ml in toluene
Formula:
C₁₈H₁₁NO₂
Description:
6-Nitrochrysene