Benzoyl peroxide 97% (dry weight) wetted with ca. 25% water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:94-36-0
:
Formula:C₁₄H₁₀O₄
MDL Number:MFCD00003071
:242.23 g/mol
Merck Index:14,1116
ADR:5.2,
:Moistened with water
EINECS:202-327-6
UN:3104
:GHS02
:GHS07
Storage Temperature:Ambient