Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methyl-2-furoic acid
3-Methyl-2-furoic acid ≥98%
3-Methyl-2-furoic acid ≥98%
:
Thermo Fisher Scientific
3-Methyl-2-furoic acid ≥98%
:
Thermo Fisher Scientific
:
4412-96-8
:
:
C₆H₆O₃
:
MFCD00014111
:
126.11 g/mol
:
133…137 °C
:
224-571-2
:
GHS07
:
Ambient
msds_ALFAL01763
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years