Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyclohexanol
Cyclohexanol
Cyclohexanol
PROOCHE105
:
LGC Standards PROMOCHEM
:
Cyclohexanol
PROOCHE105
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
2 mL
Seq:
2145
:
:
100.16 g/mol
:
13,02754
:
161 °C (1013 hPa)
:
25 °C
:
0.947 g/cm³ (20 °C)
:
59 °C
:
Ambient
:
203-630-6
:
GHS07
:
108-93-0
:
C₆H₁₂O
:
MFCD00003855