Bromocyclohexane 99%
:108-85-0
:
:C₆H₁₁Br
:MFCD00003819
:163.06 g/mol
:13,02759
:166…167 °C (760 mmHg)
:-56,5 °C
:1.33 g/cm³
:62 °C
:Ambient
:203-622-2
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4940 to 1.4960 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥98.5 %