Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Aminoquinoline
2-Aminoquinoline
2-Aminoquinoline
2-Aminoquinoline
Supplier:
Thermo Scientific
Certificates
Expand image
2-Aminoquinoline
Supplier:
Thermo Scientific
About this item
CAS Number:
580-22-3
:
Formula:
C₉H₈N₂
MDL Number:
MFCD00038079
:
144.18
Boiling Pt:
305 °C (1013 hPa)
Melting Pt:
130 °C
EINECS:
209-458-8