4-Quinolinecarbaldehyde
Catalog # MOLEM87482000
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:157.17 g/mol
Boiling Pt:122…123 °C (4 mmHg)
Melting Pt:150 °C
Storage Temperature:Refrigerator
EINECS:224-453-0
CAS Number:4363-93-3
Formula:C₁₀H₇NO
MDL Number:MFCD00006781