
4-Quinolinecarbaldehyde ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4363-93-3
:
Formula:C₁₀H₇NO
MDL Number:MFCD00006781
:157.17 g/mol
Boiling Pt:131…133 °C (5 mmHg)
Melting Pt:48…52 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:224-453-0
:GHS07